Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2,4-dimethoxyaniline, 98%, Thermo Scientific™

Catalog No. AAH5921403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5921403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5921403 Supplier Thermo Scientific Chemicals Supplier No. H5921403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-50-7
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.62
MDL Number MFCD00025752
InChI Key OLCMNCWEUMBNIS-UHFFFAOYSA-N
Synonym itr amine, benzenamine, 5-chloro-2,4-dimethoxy, 2,4-dimethoxy-5-chloroaniline, 5-chloro-2,4-dimethoxyaminobenzene, unii-lz646979la, aniline,2,4-dimethoxy-5-chloro, 5-chloro-2,4-dimethoxyphenylamine, acmc-209s8h, cambridge id 5252837, ksc496a0p
PubChem CID 66807
IUPAC Name 5-chloro-2,4-dimethoxyaniline
SMILES COC1=CC(OC)=C(Cl)C=C1N

Specifications

Melting Point 87°C to 90°C
Color Purple
Quantity 1 g
UN Number UN3077
Beilstein 2211013
Formula Weight 187.63
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Chloro-2,4-dimethoxyaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-586-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.