Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2,4-dimethoxyaniline 97.0+%, TCI America™
SDP

Catalog No. C122625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C122625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C122625G Supplier TCI America Supplier No. C122625G
Add to Cart
Add to Cart

Specifications

CAS 97-50-7
Melting Point 92°C
Color Purple
Molecular Formula C8H10ClNO2
Quantity 25 g
MDL Number MFCD00025752
Synonym itr amine, benzenamine, 5-chloro-2,4-dimethoxy, 2,4-dimethoxy-5-chloroaniline, 5-chloro-2,4-dimethoxyaminobenzene, unii-lz646979la, aniline,2,4-dimethoxy-5-chloro, 5-chloro-2,4-dimethoxyphenylamine, acmc-209s8h, cambridge id 5252837, ksc496a0p
InChI Key OLCMNCWEUMBNIS-UHFFFAOYSA-N
SMILES COC1=CC(OC)=C(Cl)C=C1N
IUPAC Name 5-chloro-2,4-dimethoxyaniline
Molecular Weight (g/mol) 187.62
PubChem CID 66807
Formula Weight 187.62
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2,4-dimethoxyaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.