Learn More
Cayman Chemical 5-Chloro-2'-deoxyuridine, Purity: ≥98%, CAS: 50-90-8, 50 mg
SDP
Catalog No. 502964882
Encompass_Preferred
A thymidine analog that is readily incorporated, following phosphorylation, into newly synthesized DNA in place of thymidine; can be detected immunologically in cells and tissues
Catalog No. 50-296-4882 Supplier Cayman Chemical Supplier No. 1815550 MG
A thymidine analog that is readily incorporated, following phosphorylation, into newly synthesized DNA in place of thymidine; can be detected immunologically in cells and tissues