Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-chloro-2-fluoro-4-[(1S,2R)-2-(2-methylpyrazol-3-yl)cyclohexyl]oxy-N-pyrimidin-4-ylbenzenesulfonamide | 1450595-86-4 | Not found on authoritative sources | 99.6% | C20H21ClFN5O3S | 10MG
SDP

Catalog No. 5000259338
Encompass_Preferred

DS-1971a is a potent, selective, and orally active NaV1.7 inhibitor used in pain research. It shows low-nanomolar inhibition of human NaV1.7, activity in rodent models, and has demonstrated analgesic effects in preclinical studies.

  • Potent NaV1.7 inhibition: hNaV1.7 IC50 = 22.8 nM; mNaV1.7 IC50 ≈ 59.4 nM.
  • Orally active with reported analgesic effects in preclinical models.
  • High purity, typically 99.6% by supplier specification.
  • Defined chemical identity: CAS 1450595-86-4; formula C20H21ClFN5O3S; MW 465.93 g·mol⁻¹.
  • Soluble in DMSO and compatible with common in vivo formulation vehicles.
  • Stable as a solid with recommended refrigerated or frozen storage for long-term retention.

Catalog No. 50-002-59338 Supplier Medchemexpress LLC Supplier No. HY13118210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DS-1971a is a potent, selective, and orally active NaV1.7 inhibitor used in pain research. It shows low-nanomolar inhibition of human NaV1.7, activity in rodent models, and has demonstrated analgesic effects in preclinical studies.

  • Potent NaV1.7 inhibition: hNaV1.7 IC50 = 22.8 nM; mNaV1.7 IC50 ≈ 59.4 nM.
  • Orally active with reported analgesic effects in preclinical models.
  • High purity, typically 99.6% by supplier specification.
  • Defined chemical identity: CAS 1450595-86-4; formula C20H21ClFN5O3S; MW 465.93 g·mol⁻¹.
  • Soluble in DMSO and compatible with common in vivo formulation vehicles.
  • Stable as a solid with recommended refrigerated or frozen storage for long-term retention.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.