Learn More
Medchemexpress LLC 5-chloro-2-fluoro-4-(hexahydropyrrolizin-8-ylmethylamino)-N-(thiazol-2-yl)benzenesulfonamide | 1788872-06-9 | 98.2% | 430.95 g/mol | C17H20ClFN4O2S2 | 10 MG
SDP
Catalog No. 5000247227
Encompass_Preferred
Nav1.7-IN-3 is a selective, orally bioavailable small-molecule inhibitor of the voltage-gated sodium channel Nav1.7 with a reported IC50 of 8 nM. The compound shows limited central nervous system penetration and is supplied for laboratory research (CAS 1788872-06-9).
  • Selective Nav1.7 inhibition (IC50 = 8 nM).
  • Orally bioavailable.
  • Limited CNS penetration.
  • High purity (~98.2%).
  • Molecular weight 430.95 g/mol; formula C17H20ClFN4O2S2.
  • Provided in small research quantities (e.g., 10 MG).
Catalog No. 50-002-47227 Supplier Medchemexpress LLC Supplier No. HY10178910MG
Nav1.7-IN-3 is a selective, orally bioavailable small-molecule inhibitor of the voltage-gated sodium channel Nav1.7 with a reported IC50 of 8 nM. The compound shows limited central nervous system penetration and is supplied for laboratory research (CAS 1788872-06-9).
  • Selective Nav1.7 inhibition (IC50 = 8 nM).
  • Orally bioavailable.
  • Limited CNS penetration.
  • High purity (~98.2%).
  • Molecular weight 430.95 g/mol; formula C17H20ClFN4O2S2.
  • Provided in small research quantities (e.g., 10 MG).