Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chloro-2-fluorophenyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. SB01971DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01971DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01971DA Supplier Thermo Scientific Chemicals Supplier No. SB01971DA
Add to Cart

Chemical Identifiers

CAS 247170-25-8
Molecular Formula C7H3ClFNS
Molecular Weight (g/mol) 187.616
MDL Number MFCD01313224
InChI Key RUASREJPMXNHLF-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorophenyl isothiocyanate, 5-chloro-2-fluorophenylisothiocyanate, 3-chloro-6-fluorophenylisothiocyanate, 5-chloro-2-fluorobenzenisothiocyanate, benzene,4-chloro-1-fluoro-2-isothiocyanato, 4-chloranyl-1-fluoranyl-2-isothiocyanato-benzene, benzene, 4-chloro-1-fluoro-2-isothiocyanato-9ci
PubChem CID 2736579
IUPAC Name 4-chloro-1-fluoro-2-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)N=C=S)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 187.62
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorophenyl isothiocyanate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.