Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C31251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31251G Supplier TCI America Supplier No. C31251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 937595-70-5
Molecular Formula C5H4BClFNO2
Molecular Weight (g/mol) 175.35
MDL Number MFCD04972408
InChI Key ZUDXVHDVUTUTFH-UHFFFAOYSA-N
Synonym 5-chloro-2-fluoropyridine-3-boronic acid, 5-chloro-2-fluoropyridin-3-yl boronic acid, 5-chloro-2-fluoropyridin-3-ylboronicacid, 2-fluoro-5-chloropyridine-3-boronic acid, 5-chloro-2-fluoro-3-pyridyl boronic acid, 5-chloro-2-fluoropyridine-3-boronic acid hydrate, pubchem17095, acmc-209rnf, 5-chloro-2-fluoro-3-pyridylboronic acid, 5-chloro-2-fluoro-3-pyridinyl boronic acid
PubChem CID 24901779
IUPAC Name (5-chloro-2-fluoropyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1F)Cl)(O)O

Specifications

Melting Point 126°C
Color White
Quantity 1 g
Formula Weight 175.35
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.