Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-chloro-2-hydroxybenzoic acid | 321-14-2 | MFCD00002457 | 98.7% | 172.57 g/mol | C7H5ClO3 | 50 G
SDP

Catalog No. 5000301017
Encompass_Preferred

5-Chloro-2-hydroxybenzoic acid is a chloro-substituted benzoic acid used as an organic reagent, analytical standard, and intermediate in life-science research. It is provided as a solid for laboratory synthesis and assay applications, with supporting documentation for safe handling and quality verification.

  • High purity suitable for analytical and synthetic use (COA available).
  • Molecular weight 172.57 g/mol.
  • Chemical formula C7H5ClO3.
  • CAS registry number 321-14-2.
  • Supplied as a solid in common laboratory pack sizes (e.g., 50 g).
  • Documentation includes SDS and batch-specific COA for traceability.

Catalog No. 50-003-01017 Supplier Medchemexpress LLC Supplier No. HYW01686650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Chloro-2-hydroxybenzoic acid is a chloro-substituted benzoic acid used as an organic reagent, analytical standard, and intermediate in life-science research. It is provided as a solid for laboratory synthesis and assay applications, with supporting documentation for safe handling and quality verification.

  • High purity suitable for analytical and synthetic use (COA available).
  • Molecular weight 172.57 g/mol.
  • Chemical formula C7H5ClO3.
  • CAS registry number 321-14-2.
  • Supplied as a solid in common laboratory pack sizes (e.g., 50 g).
  • Documentation includes SDS and batch-specific COA for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.