Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-hydroxynicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. C23391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23391G 1 g
C23395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23391G Supplier TCI America Supplier No. C23391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38076-80-1
Molecular Formula C6H3ClNO3
Molecular Weight (g/mol) 172.54
MDL Number MFCD00204158
InChI Key XBHXNMLFJZTSAS-UHFFFAOYSA-M
Synonym 5-chloro-2-hydroxynicotinic acid, 5-chloro-2-hydroxypyridine-3-carboxylic acid, 5-chloro-2-hydroxynicotinicacid, 5-chloro-2-hydroxy-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-chloro-1,2-dihydro-2-oxo, 5-chloro-2-hydroxy-nicotinic acid, 5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid, 5-chloro-2-oxo-1,2-dihydropyridine-3-carboxylic acid, maybridge1_008644, pubchem12990
PubChem CID 162286
IUPAC Name 5-chloro-2-oxo-1,2-dihydropyridine-3-carboxylate
SMILES [O-]C(=O)C1=CC(Cl)=CNC1=O

Specifications

Melting Point 262°C
Color White
Quantity 1 g
Formula Weight 173.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-hydroxynicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.