Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-iodoaniline 95.0+%, TCI America™
SDP

Catalog No. C22395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22395G Supplier TCI America Supplier No. C22395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6828-35-9
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD06738960
InChI Key FEOMAFDDLHSVMO-UHFFFAOYSA-N
Synonym benzenamine, 5-chloro-2-iodo, 5-chloro-2-iodo-phenylamine, 5-chloro-2-iodo-benzenamine, 5-chloro-2-iodophenylamine, 2-amino-4-chloro-iodobenzene, pubchem5286, 2-iodo-5-chloroaniline, 5-chloro-2-iodoanilline, 5-chloro-2-iodo-aniline, acmc-209o2s
PubChem CID 639697
IUPAC Name 5-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)N)I

Specifications

Melting Point 42°C
Quantity 5 g
Formula Weight 253.47
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-iodoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.