Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-mercaptobenzothiazole 98%, Thermo Scientific™

Catalog No. AC154910100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154910100 10 g
AC154910500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154910100 Supplier Thermo Scientific Chemicals Supplier No. 154910100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5331-91-9
Molecular Formula C7H4ClNS2
Molecular Weight (g/mol) 201.69
MDL Number MFCD00005783
InChI Key NKYDKCVZNMNZCM-UHFFFAOYSA-N
Synonym 5-chloro-2-mercaptobenzothiazole, 5-chloro-1,3-benzothiazole-2-thiol, 5-chlorobenzo d thiazole-2 3h-thione, sh-benzothiazole, 5-chloro-2-benzothiazolethiol, 2 3h-benzothiazolethione, 5-chloro, 5-chlorobenzo d thiazole-2-thiol, 5-chlorobenzothiazole-2-thiol, benzothiazole, 5-chloro-2-mercapto, 2-benzothiazolethiol, 5-chloro
PubChem CID 2723842
IUPAC Name 5-chloro-2,3-dihydro-1,3-benzothiazole-2-thione
SMILES ClC1=CC=C2SC(=S)NC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195°C to 199°C
Color Green-Yellow to Yellow
Infrared Spectrum Authentic
Quantity 10 g
Beilstein 27, II, 233
Formula Weight 201.69
Percent Purity ≥97.5%
Physical Form Granular Powder
Chemical Name or Material 5-Chloro-2-mercaptobenzothiazole, 98%
Show More Show Less
EINECSNumber 226-235-0
RTECSNumber DL6490000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.