Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. p-7028626
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2602006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2602006 Supplier Thermo Scientific Chemicals Supplier No. H2602006
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 889939-48-4
Molecular Formula C8H8Cl2O3S
Molecular Weight (g/mol) 255.109
MDL Number MFCD08445709
InChI Key BJRJHJZVSWZXIP-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxy-4-methylbenzenesulfonylchloride, 5-chloro-2-methoxy-4-methylbenzene-1-sulfonyl chloride, pubchem5095, acmc-20am4m, 5-chloro-2-methoxy-4-methylphenylsulfonyl chloride, chloro 5-chloro-2-methoxy-4-methylphenyl sulfone, 5-chloranyl-2-methoxy-4-methyl-benzenesulfonyl chloride
PubChem CID 17389862
IUPAC Name 5-chloro-2-methoxy-4-methylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C=C1Cl)S(=O)(=O)Cl)OC

Specifications

Melting Point 126°C to 128°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 255.12
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.