Learn More
eMolecules​ 419535-75-4 | 5-Chloro-2-methoxyphenylmagnesium bromide, 0.5 M in THF | Synthonix - Stock245.780 | C7H6BrClMgO | 0.000 | [Br-].COc1ccc(Cl)cc1[Mg+] | 500ml | 437237057
Catalog No. 502697728 Shop All eMolecules Inc. Products
Encompass_Preferred
5-Chloro-2-methoxyphenylmagnesium bromide, 0.5 M in THF | Synthonix - Stock | 419535-75-4245.780 | C7H6BrClMgO | 0.000 | [Br-].COc1ccc(Cl)cc1[Mg+] | 500ml | 437237057
Catalog No. 50-269-7728 Supplier eMolecules​ Supplier No. 437237057
5-Chloro-2-methoxyphenylmagnesium bromide, 0.5 M in THF | Synthonix - Stock | 419535-75-4245.780 | C7H6BrClMgO | 0.000 | [Br-].COc1ccc(Cl)cc1[Mg+] | 500ml | 437237057