Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C22925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22925G 5 g
C2292-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22925G Supplier TCI America Supplier No. C22925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89694-48-4
Molecular Formula C7H8BClO3
MDL Number MFCD01318966
InChI Key FMBVAOHFMSQDGT-UHFFFAOYSA-N
PubChem CID 2735751
IUPAC Name (5-chloro-2-methoxyphenyl)boronic acid

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 5 g
Synonym 5-chloro-2-methoxyphenyl boronic acid, 5-chloro-2-methoxybenzeneboronic acid, 5-chloro-2-methoxyphenyl boranediol, 2-methoxy-5-chlorophenylboronic acid, 4-chloroanisole-2-boronic acid, boronic acid, 5-chloro-2-methoxyphenyl, 5-chloro-2-methoxyphenylboronicacid, pubchem1782, 5-chloro-2-methoxy-phenyl boronic acid
SMILES COC1=C(C=C(Cl)C=C1)B(O)O
Molecular Weight (g/mol) 186.40
Formula Weight 186.40
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.