Learn More
eMolecules​ 1621225-24-8 | 5-Chloro-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | MFCD18712496 | 250mg
Supplier:  eMolecules​ 600850598
Encompass_Preferred
Ambeed | 4-Azatricyclo[4.3.1.138]undecan-5-one | 250mg | 600839127 | A386100 | 22607-75-6 | 165.236 | C10H15NO
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>
Catalog No. 50-213-3886