Learn More
Medchemexpress LLC 5-Chloro-2-(methylamino)benzophenone | 1022-13-5 | 98.4% | 245.71 | 100 G
SDP
Catalog No. 5000453612
Encompass_Preferred
(5-Chloro-2-(methylamino)phenyl)(phenyl)methanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Analytical reference standard categorized as a benzophenone
  • Metabolite of diazepam
  • Used as a synthetic intermediate in the synthesis of diazepam
  • Found in seized etizolam samples
  • Intended for research and forensic applications
Catalog No. 50-004-53612 Supplier Medchemexpress LLC Supplier No. HYW011991100G
(5-Chloro-2-(methylamino)phenyl)(phenyl)methanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Analytical reference standard categorized as a benzophenone
  • Metabolite of diazepam
  • Used as a synthetic intermediate in the synthesis of diazepam
  • Found in seized etizolam samples
  • Intended for research and forensic applications