Learn More
Medchemexpress LLC 5-Chloro-2-(methylamino)benzophenone | 1022-13-5 | 98.4% | 245.71 | 50 G
SDP
Catalog No. 5000452782
Encompass_Preferred
(5-Chloro-2-(methylamino)phenyl)(phenyl)methanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also an analytical reference standard categorized as a benzophenone. It is a metabolite of diazepam and has been used as a synthetic intermediate in the synthesis of diazepam. It has also been found in seized etizolam samples. This product is intended for research and forensic applications.
  • For research use only.
  • Can be used as a biological material or organic compound for life science related research.
  • Metabolite of diazepam.
  • Synthetic intermediate in the synthesis of diazepam.
  • Found in seized etizolam samples.
  • Available in various sizes.
Catalog No. 50-004-52782 Supplier Medchemexpress LLC Supplier No. HYW01199150G
(5-Chloro-2-(methylamino)phenyl)(phenyl)methanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also an analytical reference standard categorized as a benzophenone. It is a metabolite of diazepam and has been used as a synthetic intermediate in the synthesis of diazepam. It has also been found in seized etizolam samples. This product is intended for research and forensic applications.
  • For research use only.
  • Can be used as a biological material or organic compound for life science related research.
  • Metabolite of diazepam.
  • Synthetic intermediate in the synthesis of diazepam.
  • Found in seized etizolam samples.
  • Available in various sizes.