Learn More
Medchemexpress LLC 5-Chloro-2-(methylamino)benzophenone | 1022-13-5 | 98.4% | 245.71 | 500 G
SDP
Catalog No. 5000453960
Encompass_Preferred
(5-Chloro-2-(methylamino)phenyl)(phenyl)methanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research and forensic applications. 2-Methylamino-5-chlorobenzophenone is an analytical reference standard categorized as a benzophenone. It is a metabolite of diazepam and has also been used as a synthetic intermediate in the synthesis of diazepam. It has been found in seized etizolam samples.
  • Used as a biochemical reagent
  • Suitable for life science related research
  • Intended for research and forensic applications
  • Analytical reference standard
  • Metabolite of diazepam
  • Synthetic intermediate in diazepam synthesis
Catalog No. 50-004-53960 Supplier Medchemexpress LLC Supplier No. HYW011991500G
(5-Chloro-2-(methylamino)phenyl)(phenyl)methanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research and forensic applications. 2-Methylamino-5-chlorobenzophenone is an analytical reference standard categorized as a benzophenone. It is a metabolite of diazepam and has also been used as a synthetic intermediate in the synthesis of diazepam. It has been found in seized etizolam samples.
  • Used as a biochemical reagent
  • Suitable for life science related research
  • Intended for research and forensic applications
  • Analytical reference standard
  • Metabolite of diazepam
  • Synthetic intermediate in diazepam synthesis