Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Chloro-2-pentanone

Catalog No. C62603100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C62603100G 100 g
C6260325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C62603100G Supplier Sigma Aldrich Supplier No. C62603100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Chloro-2-pentanone

Specifications

CAS 5891-21-4
Density 1.057 g/mL (at 25°C (literature))
Boiling Point 71°C to 72°C (20 mmHg)
Molecular Formula C5H9ClO
Linear Formula Cl(CH2)3 COCH3
Refractive Index n20/D 1.438 (literature)
MDL Number MFCD00001008
Quantity 100 g
Synonym Methyl 3-chloropropyl ketone
Molecular Weight (g/mol) 120.58
Percent Purity 85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.