Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-pyridinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. C30001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30001G 1 g
C30005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30001G Supplier TCI America Supplier No. C30001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31181-89-2
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD08277271
InChI Key YYLBDBOSXXSZQQ-UHFFFAOYSA-N
Synonym 5-Chloro-2-formylpyridine, 5-Chloropicolinaldehyde
PubChem CID 10103267
IUPAC Name 5-chloropyridine-2-carbaldehyde
SMILES C1=CC(=NC=C1Cl)C=O

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 1 g
Formula Weight 141.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-pyridinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.