Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-thienylmagnesium bromide, 0.5M slurry in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC431970500
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111762-30-2, 109-99-9
Molecular Formula C4H2BrClMgS
Molecular Weight (g/mol) 221.78
MDL Number MFCD02260211
InChI Key BTGXLAKETDWIAB-UHFFFAOYSA-M
Synonym 5-chloro-2-thienylmagnesium bromide, magnesium, bromo 5-chloro-2-thienyl, btgxlaketdwiab-uhfffaoysa-m, bromo 5-chloro-2-thienyl magnesium, 5-chloro-2-thienyl magnesium bromide, 5-chloro-2-thienyl-magnesium bromide, 2-bromomagnesio-5-chlorothiophene, 5-chloro-2-thienylmagnesium bromide, 0.5 m slurry in thf, 5-chloro-2-thienylmagnesium bromide 0.25 m in tetrahydrofuran, 5-chloro-2-thienylmagnesium bromide, 0.25m in tetrahydrofuran, grignard reagent
PubChem CID 12345883
IUPAC Name magnesium;5-chloro-2H-thiophen-2-ide;bromide
SMILES ClC1=CC=C([Mg]Br)S1

Specifications

Color Yellow to Brown
Density 0.9690g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.969
Formula Weight 221.79
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-thienylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form expl
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.