Learn More
Accela Chembio Inc 5-chloro-2-thiophenecarboxaldehyde | 5g | 7283-96-7 | MFCD00047090 | 97+% | D: 1.376 | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4
Supplier:  Accela Chembio Inc SY0055885G
Encompass_Preferred
5-chloro-2-thiophenecarboxaldehyde | 5g | 7283-96-7 | MFCD00047090 | 97+% | D: 1.376 | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-354-352