Learn More
Aobchem 5-Chloro-3-ethoxy-2-fluorobenzaldehyde | 97% | CAS 2386740-65-2 | C9H8ClFO2 | 1g
Catalog No. 5000912868
Encompass_Preferred
5-Chloro-3-ethoxy-2-fluorobenzaldehyde | 97% | CAS 2386740-65-2 | C9H8ClFO2 | 1g
Catalog No. 50-009-12868 Supplier Aobchem Supplier No. A606761G
5-Chloro-3-ethoxy-2-fluorobenzaldehyde | 97% | CAS 2386740-65-2 | C9H8ClFO2 | 1g