Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde

Catalog No. ADE0013861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0013861G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0013861G Supplier Sigma Aldrich Supplier No. ADE0013861G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-chloro-6-iodohexane is an α,-d-dihaloalkane
  • 1,12-dichlorododecane is formed as a major product from the reduction of 1-chloro-6-iodohexane with nickel(i) salen
  • It affords 8-chloro-1-octyne on coupling with sodium acetylide.1-chloro-6-iodohexane may be used in the preparation of 1,14-dichloro-7-tetradecyne and 10-2α-(methylphenyl)-7-decynyl chloride

Specifications

Molecular Formula C8H4ClIN2O
Linear Formula C8H4ClIN2O
MDL Number MFCD20487111
Quantity 10 g
Molecular Weight (g/mol) 306.49
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.