Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-3-methylbenzo[b]thiophene 98.0+%, TCI America™
SDP

Catalog No. C245725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C245725G 25 g
C24575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C245725G Supplier TCI America Supplier No. C245725G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19404-18-3
Molecular Formula C9H7ClS
Molecular Weight (g/mol) 182.665
MDL Number MFCD00052506
InChI Key UWDQVEPXORTQFO-UHFFFAOYSA-N
Synonym 5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
PubChem CID 317317
IUPAC Name 5-chloro-3-methyl-1-benzothiophene
SMILES CC1=CSC2=C1C=C(C=C2)Cl

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 144°C
Quantity 25 g
Formula Weight 182.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.