Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-chloro-3-(thiophen-2-ylmethylene)indolin-2-one | 186611-58-5 | 98.0% | 261.73 g·mol⁻¹ | C13H8ClNOS | 10 MG
SDP

Catalog No. 5000286014
Encompass_Preferred

SU 5616 is a small-molecule research compound that modulates tyrosine kinase signal transduction and can regulate abnormal cell proliferation. It is supplied as a purified powder for laboratory in vitro research use.

  • Purity: 98.0%.
  • CAS number: 186611-58-5.
  • Molecular formula: C13H8ClNOS.
  • Molecular weight: 261.73 g·mol⁻¹.
  • Available in multiple preweighed package sizes for laboratory use.

Catalog No. 50-002-86014 Supplier Medchemexpress LLC Supplier No. HY15322710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SU 5616 is a small-molecule research compound that modulates tyrosine kinase signal transduction and can regulate abnormal cell proliferation. It is supplied as a purified powder for laboratory in vitro research use.

  • Purity: 98.0%.
  • CAS number: 186611-58-5.
  • Molecular formula: C13H8ClNOS.
  • Molecular weight: 261.73 g·mol⁻¹.
  • Available in multiple preweighed package sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.