Learn More
Apexbio Technology LLC (±)-5'-Chloro-5'-deoxy-ENBA 103626-26-2 10mg
SDP
Supplier:  Apexbio Technology LLC B743510
Encompass_Preferred
racemic Baclofen -5p-Chloro-5p-deoxy-ENBA is a small-molecule agonist targeting the adenosine A1 receptor It is designed to activate adenosine A1 receptor-mediated signaling thereby regulating neuroprotective cardioprotective and nociceptive pathways racemic Baclofen -5p-Chloro-5p-deoxy-ENBA exerts its biological activity primarily through activation of the adenosine A1 receptor In animal models administration of this compound alleviates inflammatory and neuropathic pain behaviors by modulating both peripheral and central pain pathways and influencing glial cell responses without affecting motor coordination or cardiovascular function In experimental models of Parkinson s disease it reduces L-DOPA-induced abnormal involuntary movements through modulation of striatonigral GABAergic neurotransmission Based on these pharmacological properties racemic Baclofen -5p-Chloro-5p-deoxy-ENBA holds research potential in pain management and movement disorders
Catalog No. 50-000-87807