Learn More
Cayman Chemical 5-chloro-5-DeoxyadenosIn 10g
SDP
Catalog No. 502981527
Encompass_Preferred
A nucleoside analog used as a substrate in the biosynthesis of the anticancer agent salinosporamide A
Catalog No. 50-298-1527 Supplier Cayman Chemical Supplier No. 2101210 G
A nucleoside analog used as a substrate in the biosynthesis of the anticancer agent salinosporamide A