Learn More
Cayman Chemical 5'-chloro-5'-Deoxyadenosine (hydrate), Purity: ≥98%, CAS: 698999-09-6, 1 g
SDP
Catalog No. 502983623
Encompass_Preferred
A nucleoside analog used as a substrate in the biosynthesis of the anticancer agent salinosporamide A
Catalog No. 50-298-3623 Supplier Cayman Chemical Supplier No. 210121 G
A nucleoside analog used as a substrate in the biosynthesis of the anticancer agent salinosporamide A