Learn More
AdipoGen 5-Chloro-5-deoxyadenosine
SDP
Supplier:  AdipoGen CDXC0345M250
Encompass_Preferred
Chemical. CAS 892-48-8. Formula C10H12ClN5O3. MW 285.7. Synthetic. Nucleoside derivative used as a synthesis intermediate. Adenosine agonist. Inhibits phosphorylation of troponin I in rat heart and phosphatidylinositol and myosin light chain in calf aorta smooth muscle.
Catalog No. 50-256-1373