Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Chloro-7-azaindole | 866546-07-8 | MFCD06659676 | 99.9% | 152.58 | 1 ML
SDP

Catalog No. 5000404131
Encompass_Preferred

5-Chloro-7-azaindole is a chemical compound primarily utilized as an organic synthesis intermediate and a pharmaceutical intermediate. It finds application in laboratory research and development, as well as in chemical and pharmaceutical production processes.

  • Off-white to light yellow solid
  • Purity of 99.9% (HPLC)
  • Melting point: 161-162°C
  • Density: 1.425±0.06 g/cm³ (predicted)
  • Store in a dark, dry place at room temperature
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at -80°C in solvent

Catalog No. 50-004-04131 Supplier Medchemexpress LLC Supplier No. HY3001110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Chloro-7-azaindole is a chemical compound primarily utilized as an organic synthesis intermediate and a pharmaceutical intermediate. It finds application in laboratory research and development, as well as in chemical and pharmaceutical production processes.

  • Off-white to light yellow solid
  • Purity of 99.9% (HPLC)
  • Melting point: 161-162°C
  • Density: 1.425±0.06 g/cm³ (predicted)
  • Store in a dark, dry place at room temperature
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at -80°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.