Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Chloro-7-iodo-8-quinolinol

Catalog No. 24880500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-880-500G 500 g
24-880-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-880-500G Supplier Sigma Aldrich Supplier No. 24880500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-chloro-7-iodo-8-quinolinol is an antibiotic with metal-binding properties and exhibits anticancer activity in vitro and in vivo
  • 5-chloro-7-iodo-8-quinolinol, its glucuronide and sulfate in serum, urine and milk has been quantitated by gas chromatography.5-chloro-7-iodo-8-quinolinol has been used in analysis of intracellular free zinc ions with the fluozin-3 probes using fluorescence microscopy

Specifications

CAS 130-26-7
Molecular Formula C9H5ClINO
Linear Formula C9H5ClINO
MDL Number MFCD00006787
Quantity 500 g
Synonym 5-Chloro-8-hydroxy-7-iodoquinoline; Clioquinol; Iodochlorhydroxyquin
Molecular Weight (g/mol) 305.5
Percent Purity ≥95.0% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.