Learn More
Cayman Chemical 5-chloro AB-PINACA 10mg
SDP
Catalog No. 502894144
Encompass_Preferred
A derivative of AB-PINACA featuring a chlorine atom added to the terminal carbon of the pentyl group; intended for research and forensic applications
Catalog No. 50-289-4144 Supplier Cayman Chemical Supplier No. 900185710 MG
A derivative of AB-PINACA featuring a chlorine atom added to the terminal carbon of the pentyl group; intended for research and forensic applications