Learn More
Cayman Chemical 5-chloro AB-PINACA, Purity: ≥98%, CAS: 1801552-02-2, 1 mg
SDP
Catalog No. 502894576
Encompass_Preferred
A derivative of AB-PINACA featuring a chlorine atom added to the terminal carbon of the pentyl group; intended for research and forensic applications
Catalog No. 50-289-4576 Supplier Cayman Chemical Supplier No. 90018571 MG
A derivative of AB-PINACA featuring a chlorine atom added to the terminal carbon of the pentyl group; intended for research and forensic applications