Learn More
Cayman Chemical 5-chloro AB-PINACA 5mg
SDP
Catalog No. 502893116
Encompass_Preferred
A derivative of AB-PINACA featuring a chlorine atom added to the terminal carbon of the pentyl group; intended for research and forensic applications
Catalog No. 50-289-3116 Supplier Cayman Chemical Supplier No. 90018575 MG
A derivative of AB-PINACA featuring a chlorine atom added to the terminal carbon of the pentyl group; intended for research and forensic applications