Learn More
Medchemexpress LLC 5-chloro-n-[2-(4-hydroxysulfamoyl-phenyl)-ethyl]-2-methoxy-benzamide | 2112809-98-8 | 98.4% | 384.83 | C16H17ClN2O5S | 100 MG
SDP
Catalog No. 5000228145
Encompass_Preferred
JC-171 is a small-molecule research compound that selectively inhibits the NLRP3 inflammasome and reduces LPS/ATP-induced interleukin-1β (IL-1β) release from J774A.1 macrophages (IC50 8.45 μM). It is supplied as a high-purity powder for laboratory research and in vivo studies.
  • Selective NLRP3 inflammasome inhibitor.
  • Inhibits LPS/ATP-induced IL-1β release (IC50 8.45 μM).
  • High purity (98.4%) and defined molecular weight (384.83).
  • Recommended storage: powder stability at -20°C (3 years) and 4°C (2 years).
  • Supplied as a 100 MG powder for research use.
Catalog No. 50-002-28145 Supplier Medchemexpress LLC Supplier No. HY117432100MG
JC-171 is a small-molecule research compound that selectively inhibits the NLRP3 inflammasome and reduces LPS/ATP-induced interleukin-1β (IL-1β) release from J774A.1 macrophages (IC50 8.45 μM). It is supplied as a high-purity powder for laboratory research and in vivo studies.
  • Selective NLRP3 inflammasome inhibitor.
  • Inhibits LPS/ATP-induced IL-1β release (IC50 8.45 μM).
  • High purity (98.4%) and defined molecular weight (384.83).
  • Recommended storage: powder stability at -20°C (3 years) and 4°C (2 years).
  • Supplied as a 100 MG powder for research use.