Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-chloro-N-[2-(4-hydroxysulfamoyl-phenyl)-ethyl]-2-methoxy-benzamide | 2112809-98-8 | 98.4% | 384.83 g/mol | C16H17ClN2O5S | 50 MG
SDP

Catalog No. 5000226690
Encompass_Preferred

JC-171 is a selective NLRP3 inflammasome inhibitor that reduces LPS/ATP-induced interleukin-1β (IL-1β) release from J774A.1 macrophages with an IC50 of 8.45 μM. It is supplied as a solid and as a DMSO solution for research use, with reported high purity and defined storage recommendations.

  • Selective NLRP3 inflammasome inhibition in cell-based assays.
  • Activity: IC50 8.45 μM for inhibition of IL-1β release.
  • High reported purity: 98.41%.
  • Molecular weight 384.83 g/mol; formula C16H17ClN2O5S.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • Storage recommendations provided for powder and in-solution stability.

Catalog No. 50-002-26690 Supplier Medchemexpress LLC Supplier No. HY11743250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JC-171 is a selective NLRP3 inflammasome inhibitor that reduces LPS/ATP-induced interleukin-1β (IL-1β) release from J774A.1 macrophages with an IC50 of 8.45 μM. It is supplied as a solid and as a DMSO solution for research use, with reported high purity and defined storage recommendations.

  • Selective NLRP3 inflammasome inhibition in cell-based assays.
  • Activity: IC50 8.45 μM for inhibition of IL-1β release.
  • High reported purity: 98.41%.
  • Molecular weight 384.83 g/mol; formula C16H17ClN2O5S.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • Storage recommendations provided for powder and in-solution stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.