Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-chloro-N-[5-chloro-4-[(4-chlorophenyl)(cyano)methyl]-2-methylphenyl]-2-hydroxybenzamide | 212135-62-1 | 96.0% | C22H15Cl3N2O2 | 10MG
SDP

Catalog No. 5000261216
Encompass_Preferred

ZT-1a is a small-molecule, non-ATP-competitive inhibitor of STE20/SPS1-related proline-alanine-rich kinase (SPAK/STK39). It has been reported to inhibit SPAK enzymatic activity with mid-micromolar IC50 values and is used as a biochemical probe to study SPAK-dependent regulation of ion transporters.

  • Non-ATP-competitive inhibition of SPAK
  • Reported IC50 values: 44.3 μM (0.01 mM ATP), 35.0 μM (0.1 mM ATP), 46.7 μM (1 mM ATP)
  • Molecular formula: C22H15Cl3N2O2
  • Molecular weight: 445.73 g·mol⁻¹
  • Useful as a biochemical probe for studying SPAK-dependent signaling
  • Available in small research quantities for in vitro studies

Catalog No. 50-002-61216 Supplier Medchemexpress LLC Supplier No. HY13653210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZT-1a is a small-molecule, non-ATP-competitive inhibitor of STE20/SPS1-related proline-alanine-rich kinase (SPAK/STK39). It has been reported to inhibit SPAK enzymatic activity with mid-micromolar IC50 values and is used as a biochemical probe to study SPAK-dependent regulation of ion transporters.

  • Non-ATP-competitive inhibition of SPAK
  • Reported IC50 values: 44.3 μM (0.01 mM ATP), 35.0 μM (0.1 mM ATP), 46.7 μM (1 mM ATP)
  • Molecular formula: C22H15Cl3N2O2
  • Molecular weight: 445.73 g·mol⁻¹
  • Useful as a biochemical probe for studying SPAK-dependent signaling
  • Available in small research quantities for in vitro studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.