Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-chloro-N,N,2-trimethyl-1H-indole-3-ethanamine | 1210-81-7 | MFCD22201006 | 99.7% | 236.74 | C13H17ClN2 | 100 MG
SDP

Catalog No. 5000247182
Encompass_Preferred

ST1936 (CAS 1210-81-7) is a small-molecule research compound reported as a selective 5-HT6 receptor agonist. It is supplied as a solid with high purity, and is characterized by molecular formula C13H17ClN2 and molecular weight 236.74 g/mol.

  • Research compound acting as a selective 5-HT6 receptor agonist.
  • High purity: 99.7%.
  • Chemical formula C13H17ClN2; molecular weight 236.74 g/mol.
  • Appearance: solid, white to light yellow.
  • Supplied quantity: 100 mg.
  • Storage (powder): -20°C up to 3 years; 4°C up to 2 years.
  • Storage (in solvent): -80°C up to 6 months; -20°C up to 1 month.
  • Shipping: room temperature in continental US; may vary elsewhere.

Catalog No. 50-002-47182 Supplier Medchemexpress LLC Supplier No. HY103110100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ST1936 (CAS 1210-81-7) is a small-molecule research compound reported as a selective 5-HT6 receptor agonist. It is supplied as a solid with high purity, and is characterized by molecular formula C13H17ClN2 and molecular weight 236.74 g/mol.

  • Research compound acting as a selective 5-HT6 receptor agonist.
  • High purity: 99.7%.
  • Chemical formula C13H17ClN2; molecular weight 236.74 g/mol.
  • Appearance: solid, white to light yellow.
  • Supplied quantity: 100 mg.
  • Storage (powder): -20°C up to 3 years; 4°C up to 2 years.
  • Storage (in solvent): -80°C up to 6 months; -20°C up to 1 month.
  • Shipping: room temperature in continental US; may vary elsewhere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.