Learn More
Cayman Chemical 5-chloro NNEI, Purity: ≥98%, CAS: 1800101-23-8, 1 mg
SDP
Catalog No. 502969645
Encompass_Preferred
A derivative of NNEI that has a chloride atom added to the terminal carbon of the pentyl group; intended for forensic and research applications
Catalog No. 50-296-9645 Supplier Cayman Chemical Supplier No. 160031 MG
A derivative of NNEI that has a chloride atom added to the terminal carbon of the pentyl group; intended for forensic and research applications