Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-o-anisic acid, 99%, Thermo Scientific™

Catalog No. AC393461000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC393461000 Supplier Thermo Scientific Chemicals Supplier No. 393461000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3438-16-2
MDL Number MFCD00017546
InChI Key HULDRQRKKXRXBI-UHFFFAOYSA-N
Synonym 5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid
PubChem CID 76970
IUPAC Name 5-chloro-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)Cl)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 96°C to 99°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7ClO3
Linear Formula CH3OC6H3(Cl)CO2H
Quantity 100 g
Molecular Weight (g/mol) 186.591
Formula Weight 186.59
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-o-anisic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.