missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorofuro[3,2-b]pyridine, 96%, Thermo Scientific™

Catalog No. AAH3309903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3309903 1 g
AAH33099MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3309903 Supplier Thermo Scientific Chemicals Supplier No. H3309903

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As a potent antiallergy agents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182691-76-5
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD15143869
InChI Key LBJDELVHPFXIRO-UHFFFAOYSA-N
Synonym 5-chlorofuro 3,2-b pyridine, furo 3,2-b pyridine, 5-chloro
PubChem CID 10606968
IUPAC Name 5-chlorofuro[3,2-b]pyridine
SMILES ClC1=CC=C2OC=CC2=N1

Specifications

Melting Point 50°C to 53°C
Quantity 1 g
Formula Weight 153.57
Percent Purity 96%
Chemical Name or Material 5-Chlorofuro[3,2-b]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.