Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Chloromethyl-1-propyl-1H-imidazole hydrochloride | 497223-15-1 | 95.0% | 195.09 | 500 MG
SDP

Catalog No. 5000404012
Encompass_Preferred

5-Chloromethyl-1-propyl-1H-imidazole hydrochloride is a drug intermediate for synthesis of various active compounds.

  • Drug intermediate for synthesis of various active compounds.
  • Available in various quantities (50 mg, 100 mg, 250 mg, 500 mg, 1 g).
  • Estimated time of arrival: April 10.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
  • For research use only.

Catalog No. 50-004-04012 Supplier Medchemexpress LLC Supplier No. HY42850500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Chloromethyl-1-propyl-1H-imidazole hydrochloride is a drug intermediate for synthesis of various active compounds.

  • Drug intermediate for synthesis of various active compounds.
  • Available in various quantities (50 mg, 100 mg, 250 mg, 500 mg, 1 g).
  • Estimated time of arrival: April 10.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.