Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloromethyl-2-oxazolidinone 98.0+%, TCI America™
SDP

Catalog No. C12565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12565G Supplier TCI America Supplier No. C12565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heterocyclic building block for chemical synthesis.

Chemical Identifiers

CAS 22625-57-6
Molecular Formula C4H6ClNO2
Molecular Weight (g/mol) 135.547
MDL Number MFCD00005269
InChI Key FNOZCEQRXKPZEZ-UHFFFAOYSA-N
Synonym 5-chloromethyl-2-oxazolidinone, 5-chloromethyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 5-chloromethyl, 5-chloromethyl oxazolidin-2-one, 5-chloromethyloxazolidin-2-one, 2-oxazolidinone,5-chloromethyl, acmc-209fxq, fnozceqrxkpzez-uhfffaoysa, 5-chloromethyl-1,3-oxazolidin-2-one #
PubChem CID 90815
IUPAC Name 5-(chloromethyl)-1,3-oxazolidin-2-one
SMILES C1C(OC(=O)N1)CCl

Specifications

Melting Point 105°C
Color White
Quantity 5 g
Formula Weight 135.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloromethyl-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.