Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Chloromethyl)-2-phenylpyrimidine, 97%, Thermo Scientific™

Catalog No. CC66324DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC66324DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC66324DA Supplier Thermo Scientific Chemicals Supplier No. CC66324DA
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 886531-63-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H9ClN2
Quantity 1 g
MDL Number MFCD09879948
Synonym 5-chloromethyl-2-phenylpyrimidine, 5-chloromethyl pyrimidin-2-yl benzene, 5-chloromethyl-2-phenyl-1,3-diazine, pyrimidine, 5-chloromethyl-2-phenyl, pyrimidine, 5-chloromethyl-2-phenyl-9ci
InChI Key VJINEZQXTOUKNJ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=NC=C(C=N2)CCl
IUPAC Name 5-(chloromethyl)-2-phenylpyrimidine
Molecular Weight (g/mol) 204.657
PubChem CID 24229726
Formula Weight 204.66
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-2-phenylpyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.