Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloroquinoline, Thermo Scientific™

Catalog No. AC469740010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469740010 1 g
AC469740050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469740010 Supplier Thermo Scientific Chemicals Supplier No. 469740010
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material 5-Chloroquinoline
Alpha Vector CHLOROQUINOLINE
CAS 635-27-8
Melting Point 28°C to 32°C
Flash Point 263°C
Packaging Glass bottle
Molecular Formula C9 H6 Cl N
Quantity 1 g
MDL Number 00239413
Molecular Weight (g/mol) 163.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.