Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Chlorovaleroyl chloride

Catalog No. 12524550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-524-550G 50 g
12-524-510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-524-550G Supplier Sigma Aldrich Supplier No. 12524550G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-chlorovaleroyl chloride is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.5-chlorovaleroyl chloride (5-cvc) has been used in the development and validation of a GC-fid method for the analysis of low level impurities present in 5-cvc
  • It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine

Specifications

CAS 1575-61-7
Density 1.2 g/mL (at 25°C (literature))
Boiling Point 38°C to 39°C (0.15 mmHg)
Molecular Formula C5H8Cl2O
Refractive Index n20/D 1.464 (literature)
Linear Formula Cl(CH2)4 COCl
MDL Number MFCD00000758
Quantity 50 g
Synonym 5-Chloropentanoyl chloride
Molecular Weight (g/mol) 155.02
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.