Learn More
Medchemexpress LLC 5α-cholestan-3β-ol | 80-97-7 | MFCD00066413 | 99.9% | 388.67 g/mol | C27H48O | 50 MG
SDP
Catalog No. 5000244488
Encompass_Preferred
5α-Cholestan-3β-ol (dihydrocholesterol) is a derivatized steroid compound used as an analytical standard and research reagent in lipid analysis. It is derived from cholesterol and is provided as a high-purity solid for chromatographic and analytical workflows.
  • Derivatized steroid compound derived from cholesterol.
  • High purity (99.9%) suitable for analytical applications.
  • Suitable as a standard in HPLC lipid analysis.
  • Synonyms include dihydrocholesterol and 5α-cholestanol.
  • Molecular formula C27H48O and molecular weight 388.67 g/mol.
Catalog No. 50-002-44488 Supplier Medchemexpress LLC Supplier No. HY10781950MG
5α-Cholestan-3β-ol (dihydrocholesterol) is a derivatized steroid compound used as an analytical standard and research reagent in lipid analysis. It is derived from cholesterol and is provided as a high-purity solid for chromatographic and analytical workflows.
  • Derivatized steroid compound derived from cholesterol.
  • High purity (99.9%) suitable for analytical applications.
  • Suitable as a standard in HPLC lipid analysis.
  • Synonyms include dihydrocholesterol and 5α-cholestanol.
  • Molecular formula C27H48O and molecular weight 388.67 g/mol.