Learn More
Medchemexpress LLC 5α-cholestan-3β-ol-d5 | 2260669-14-3 | 99.2% | 393.70 g·mol⁻¹ | C27H43D5O | 5 MG
SDP
Catalog No. 5000242796
Encompass_Preferred
5α-Cholestan-3β-ol-d5 is the deuterium-labeled analogue of 5α-cholestan-3β-ol (dihydrocholesterol). It is provided as a stable-isotope internal standard for analytical and biochemical applications, designed for accurate quantitation of sterols in mass spectrometry and isotope-dilution workflows.
  • Stable isotope-labeled internal standard for quantitative mass spectrometry
  • High purity (99.2%) suitable for analytical workflows
  • Suitable for isotope-dilution experiments and method validation
  • Available in small mg-scale quantities for trace analysis
  • Molecular formula C27H43D5O; molecular weight 393.70 g·mol⁻¹
Catalog No. 50-002-42796 Supplier Medchemexpress LLC Supplier No. HY107819S5MG
5α-Cholestan-3β-ol-d5 is the deuterium-labeled analogue of 5α-cholestan-3β-ol (dihydrocholesterol). It is provided as a stable-isotope internal standard for analytical and biochemical applications, designed for accurate quantitation of sterols in mass spectrometry and isotope-dilution workflows.
  • Stable isotope-labeled internal standard for quantitative mass spectrometry
  • High purity (99.2%) suitable for analytical workflows
  • Suitable for isotope-dilution experiments and method validation
  • Available in small mg-scale quantities for trace analysis
  • Molecular formula C27H43D5O; molecular weight 393.70 g·mol⁻¹