Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5α-cholestan-3-one | 566-88-1 | MFCD00065901 | 99.3% | 386.65 g/mol | C27H46O | 1 ML
SDP

Catalog No. 5000244515
Encompass_Preferred

5α-Cholestan-3-one is an endogenous steroid metabolite supplied as a solid and as a ready-to-use 10 mM solution in ethanol. It is used in biochemical and pharmacological studies of sterol metabolism and related pathways. High purity and multiple package formats support research workflows.

  • High purity: 99.3%.
  • Available as a solid and a 10 mM solution in ethanol.
  • Soluble in ethanol at 14.29 mg/mL; may require ultrasonic agitation.
  • Molecular formula C27H46O, molecular weight 386.65 g/mol.
  • Recommended storage: powder -20°C for long term, solution -80°C for extended storage.
  • Suitable for biochemical and pharmacological research involving sterol metabolism.

Catalog No. 50-002-44515 Supplier Medchemexpress LLC Supplier No. HY10782610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5α-Cholestan-3-one is an endogenous steroid metabolite supplied as a solid and as a ready-to-use 10 mM solution in ethanol. It is used in biochemical and pharmacological studies of sterol metabolism and related pathways. High purity and multiple package formats support research workflows.

  • High purity: 99.3%.
  • Available as a solid and a 10 mM solution in ethanol.
  • Soluble in ethanol at 14.29 mg/mL; may require ultrasonic agitation.
  • Molecular formula C27H46O, molecular weight 386.65 g/mol.
  • Recommended storage: powder -20°C for long term, solution -80°C for extended storage.
  • Suitable for biochemical and pharmacological research involving sterol metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.